Learn More
MSC Ashland Conveyor Replacement Roller, 1.9 in, 7/16 in, Galvanized Steel
Catalog No. 5000513722
Encompass
This replacement conveyor roller is designed for use in conveyor systems requiring a 21-7/8 in between frame dimension. It features a 1.9 in diameter and a 7/16 in axle, constructed from 14 gage galvanized steel for durability.
  • 1.9 in diameter
  • 7/16 in axle
  • Constructed from 14 gage galvanized steel
  • Designed for conveyor systems with 21-7/8 in between frames
Catalog No. 50-005-13722 Supplier MSC Supplier No. 66065103
This replacement conveyor roller is designed for use in conveyor systems requiring a 21-7/8 in between frame dimension. It features a 1.9 in diameter and a 7/16 in axle, constructed from 14 gage galvanized steel for durability.
  • 1.9 in diameter
  • 7/16 in axle
  • Constructed from 14 gage galvanized steel
  • Designed for conveyor systems with 21-7/8 in between frames