Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ashton Pumpmatic PUMPMATIC™ Liquid-Dispensing System
SDP

Catalog No. 22246661
Encompass
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
Sterility:
Non-sterile
Packaging Type:
Bulk
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Packaging Type
22-246661 1 mL Non-sterile Bulk
22-246662 5 mL Non-sterile Bulk
22-246663 10 mL Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-246661 Supplier Ashton Pumpmatic Supplier No. ASHTON201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid, drop-free fluid sampling, transferring, and dispensing

  • Speeds and simplifies fluid sampling — from 1mL to 10mL per sample
  • Serological pipette/dispenser combination offers accuracy of a reusable at the cost of a disposable
  • Controlled rate of sampling and dispensing delivers accurate volumes without drips or spills
  • Hazardous liquids do not need to be closer than arm's length
  • Transparent, totally closed system
  • Features accurate, clearly marked, easily-read graduations
  • Smooth-action syringe and plunger with Luer-Lok™ permit rapid, one-handed sampling and dispensing
  • Lightweight, unbreakable, clear polystyrene

Specifications

Material Polystyrene
Sterility Non-sterile
Packaging Type Bulk
Volume (English) 0.03 oz.
Volume (Metric) 1 mL
Type Sampling/Dispensing Syringe
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.