Learn More
Alkali Scientific ASI™ Fetal Bovine Serum (FBS), Heat Inactivated, USDA Approved, 500mL
SDP
Catalog No. 502335984
Encompass_Preferred
Alkali Scientific™ Premium Imported (USDA Approved) FBS is ideal for the most sensitive cell lines and can be used to culture all cell types including macrophages, cancer and cancer related cells, Hybridomas and more. It works great for all our human primary cells and stem cells. Our Alkali Scientific™ FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance. Triple 0.1µm sterile-filtered. Mycoplasma-tested and virus-screened. Ships on dry ice. Store frozen at -20 to -10°C.
Catalog No. 50-233-5984 Supplier Alkali Scientific Supplier No. FB73
Alkali Scientific™ Premium Imported (USDA Approved) FBS is ideal for the most sensitive cell lines and can be used to culture all cell types including macrophages, cancer and cancer related cells, Hybridomas and more. It works great for all our human primary cells and stem cells. Our Alkali Scientific™ FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance. Triple 0.1µm sterile-filtered. Mycoplasma-tested and virus-screened. Ships on dry ice. Store frozen at -20 to -10°C.