Learn More
Alkali Scientific ASI™ Fetal Bovine Serum (FBS), US Source, 500mL
SDP
Catalog No. 502336102
Encompass_Preferred
Alkali Scientific™ U.S. Source Fetal Bovine Sera is collected entirely from USDA licensed abattoirs and processed within the United States. Our Alkali Scientific™ serum is maintained at or below 40°F from collection through sterile bottling to ensure superior performance. It meets the highest industry standards for endotoxin and hemoglobin levels and supports the growth of the widest variety of cell lines. Our Premium US Origin FBS is ideal for the most sensitive cell lines and can be used to culture all cell types including macrophages, cancer and cancer related cells, Hybridomas and more. Triple 0.1µm sterile-filtered. Mycoplasma-tested and virus-screened. Ships on dry ice. Store frozen at -20 to -10°C.
Catalog No. 50-233-6102 Supplier Alkali Scientific Supplier No. FB60
Alkali Scientific™ U.S. Source Fetal Bovine Sera is collected entirely from USDA licensed abattoirs and processed within the United States. Our Alkali Scientific™ serum is maintained at or below 40°F from collection through sterile bottling to ensure superior performance. It meets the highest industry standards for endotoxin and hemoglobin levels and supports the growth of the widest variety of cell lines. Our Premium US Origin FBS is ideal for the most sensitive cell lines and can be used to culture all cell types including macrophages, cancer and cancer related cells, Hybridomas and more. Triple 0.1µm sterile-filtered. Mycoplasma-tested and virus-screened. Ships on dry ice. Store frozen at -20 to -10°C.