Learn More
Alkali Scientific ASI™ Fetal Bovine Serum (FBS), USDA Approved, 500 mL
SDP
Catalog No. 502336103
Encompass_Preferred
Alkali Scientific™ Fetal Bovine Serum (FBS) is the liquid part of the blood depleted of the cellular fraction and coagulation factors. It is extracted after centrifugation of coagulated fetal bovine blood collected aseptically. Sterile FBS is obtained by triple filtration of 0.1µm. FBS contains proteins and growth factors allowing maintenance and proliferation of eukaryote cells in vitro. It also contains attachment factors that create a “coating” effect on the plastic of the support, thus increasing cell adhesion. It is an essential product in cell culture because of the different components it provides. All Alkali Scientific™ FBS batches are tested for endotoxin and mycoplasma levels, specific viruses testing (BVD), hemoglobin and total protein rates and cell culture performance. Triple 0.1µm sterile-filtered. Mycoplasma-tested and virus-screened. Ships on dry ice. Store frozen at -20 to -10°C.
Catalog No. 50-233-6103 Supplier Alkali Scientific Supplier No. FB72
Alkali Scientific™ Fetal Bovine Serum (FBS) is the liquid part of the blood depleted of the cellular fraction and coagulation factors. It is extracted after centrifugation of coagulated fetal bovine blood collected aseptically. Sterile FBS is obtained by triple filtration of 0.1µm. FBS contains proteins and growth factors allowing maintenance and proliferation of eukaryote cells in vitro. It also contains attachment factors that create a “coating” effect on the plastic of the support, thus increasing cell adhesion. It is an essential product in cell culture because of the different components it provides. All Alkali Scientific™ FBS batches are tested for endotoxin and mycoplasma levels, specific viruses testing (BVD), hemoglobin and total protein rates and cell culture performance. Triple 0.1µm sterile-filtered. Mycoplasma-tested and virus-screened. Ships on dry ice. Store frozen at -20 to -10°C.