Learn More
Alkali Scientific ASI™ Gentamicin Sulfate Solution (10 mg/mL), 6 x 20mL
SDP
Catalog No. 502389557
Encompass_Preferred
This wide range of antibiotics is ideal for the prevention or control of microbial contaminants in cell culture applications. Antibiotics prevent the overgrowth of bacteria, yeast, fungi, or mycoplasma which can adversely affect cell growth. Low toxicity, broad antimicrobial spectrum, minimizing the formation of resistant microorganisms, and eliminating contamination should all be considered when selecting an antibiotic. Antibiotics should be used in a limited and controlled environment as they can suppress the level of contamination so that it is undetectable and cause certain microorganisms to become resistant.An economical alternative to liquid media, this complete line of powdered media comes in 10- and 50-L sizes. Dry media is supplied in plastic bottles. Shipped at ambient temperature. Recommended storage 2 to 8°C (36 to 46°F).
Catalog No. 50-238-9557 Supplier Alkali Scientific Supplier No. FB92
This wide range of antibiotics is ideal for the prevention or control of microbial contaminants in cell culture applications. Antibiotics prevent the overgrowth of bacteria, yeast, fungi, or mycoplasma which can adversely affect cell growth. Low toxicity, broad antimicrobial spectrum, minimizing the formation of resistant microorganisms, and eliminating contamination should all be considered when selecting an antibiotic. Antibiotics should be used in a limited and controlled environment as they can suppress the level of contamination so that it is undetectable and cause certain microorganisms to become resistant.An economical alternative to liquid media, this complete line of powdered media comes in 10- and 50-L sizes. Dry media is supplied in plastic bottles. Shipped at ambient temperature. Recommended storage 2 to 8°C (36 to 46°F).