Learn More
Alkali Scientific ASI™ Media Storage Bottle, 1000mL with Standard (GL45) Blue Cap, 10/cs
SDP
Catalog No. 502336145
Encompass_Preferred
Hybex reusable glass media bottles are manufactured from high quality, borosilicate 3.3 glass. With their superior chemical resistance, these bottles are ideal for storage of reagents, culture media, biological fluids and a variety of other aqueous and nonaqueous solutions. A low coefficient of thermal expansion (3.3), provides temperature resistance that is far superior to that of standard lab glass (or soda lime glass). Hybex glass can withstand temperatures from -70 to 500°C, permitting repeated use in freezers, microwaves, autoclaves and dry heat sterilization chambers. All hybex media bottles include a drip-free sealing ring as well as a standard blue polypropylene (GL45) sealing cap. Ten per pack.
Catalog No. 50-233-6145 Supplier Alkali Scientific Supplier No. RS70205B
Hybex reusable glass media bottles are manufactured from high quality, borosilicate 3.3 glass. With their superior chemical resistance, these bottles are ideal for storage of reagents, culture media, biological fluids and a variety of other aqueous and nonaqueous solutions. A low coefficient of thermal expansion (3.3), provides temperature resistance that is far superior to that of standard lab glass (or soda lime glass). Hybex glass can withstand temperatures from -70 to 500°C, permitting repeated use in freezers, microwaves, autoclaves and dry heat sterilization chambers. All hybex media bottles include a drip-free sealing ring as well as a standard blue polypropylene (GL45) sealing cap. Ten per pack.