Learn More
Alkali Scientific ASI™ Solution Reservoir, 10mL, Sterile, Individually Wrapped, 100/cs
SDP
Catalog No. 502336115
Encompass_Preferred
ASI Solution Reservoirs are designed for use with all multichannel pipettes. Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented “spouts” in each corner. Bottoms are formed with a sharply tapered “V” shape to minimize waste and reagent consumption. Easy open, hassle free white tear strips are featured for easy handling. Stacked basins will not "stick" to each other. Gamma irradiated sterile certified RNase, DNase, pyrogen free. 100 per pack.
Catalog No. 50-233-6115 Supplier Alkali Scientific Supplier No. RS711
ASI Solution Reservoirs are designed for use with all multichannel pipettes. Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented “spouts” in each corner. Bottoms are formed with a sharply tapered “V” shape to minimize waste and reagent consumption. Easy open, hassle free white tear strips are featured for easy handling. Stacked basins will not "stick" to each other. Gamma irradiated sterile certified RNase, DNase, pyrogen free. 100 per pack.