Learn More
Alkali Scientific ASI™ Spheroid Forming Gel, Sterile, 5/Cs
SDP
Catalog No. 5000662756
Encompass_Preferred
Spheroid Forming Gel is designed to support the formation of uniform spheroids and is well suited for advanced 3D cell culture applications. The gel provides a supportive microenvironment that promotes cell aggregation and stable spheroid growth, making it useful for cancer research, drug screening, tissue engineering, and stem cell studies. Its optimized formulation allows easy handling and simplifies the process of collecting formed spheroids without damaging delicate cell structures. The material is based on hyaluronic acid, a biocompatible component that helps maintain cell viability and structural integrity during culture. The gel swells to approximately 280 μm, providing a consistent environment that supports reproducible spheroid and formation. Each package contains a total of 1 g of gel supplied as 0.2 g per tube, allowing convenient preparation and use in laboratory workflows.
Catalog No. 50-006-62756 Supplier Alkali Scientific Supplier No. SFG05
Add to Cart
Add to Cart
Spheroid Forming Gel is designed to support the formation of uniform spheroids and is well suited for advanced 3D cell culture applications. The gel provides a supportive microenvironment that promotes cell aggregation and stable spheroid growth, making it useful for cancer research, drug screening, tissue engineering, and stem cell studies. Its optimized formulation allows easy handling and simplifies the process of collecting formed spheroids without damaging delicate cell structures. The material is based on hyaluronic acid, a biocompatible component that helps maintain cell viability and structural integrity during culture. The gel swells to approximately 280 μm, providing a consistent environment that supports reproducible spheroid and formation. Each package contains a total of 1 g of gel supplied as 0.2 g per tube, allowing convenient preparation and use in laboratory workflows.