Learn More
Alkali Scientific ASI Supreme™ Frosted Plus Charged Glass Microscope Slides, Blue Writing Area, 20 Inner Packs of 72, 1440/cs
SDP
Catalog No. 502335996
Encompass_Preferred
ASI Supreme™ Frosted Plus Charged Glass slides have a positive electrical charge and crystal clear glass. They are ideal for pathology, tumor sectioning and cell imaging. They are validated on Thermo Scientific SlideMate/SlideMate AS and other automated inkjet printers.
Catalog No. 50-233-5996 Supplier Alkali Scientific Supplier No. SM2575B
ASI Supreme™ Frosted Plus Charged Glass slides have a positive electrical charge and crystal clear glass. They are ideal for pathology, tumor sectioning and cell imaging. They are validated on Thermo Scientific SlideMate/SlideMate AS and other automated inkjet printers.