Learn More
Alkali Scientific ASI Supreme™ Frosted Plus Charged Glass Microscope Slides, 20 Inner Packs of 72, 1440/cs
SDP
Catalog No. 502335819
Encompass_Preferred
ASI™ Supreme Frosted Glass Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched. These microscope slides are positively charged, to help in holding thin or powdery samples and make them adhere to the slide. This makes the mounting process easier and makes the viewing more successful.
Catalog No. 50-233-5819 Supplier Alkali Scientific Supplier No. SM2575
ASI™ Supreme Frosted Glass Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched. These microscope slides are positively charged, to help in holding thin or powdery samples and make them adhere to the slide. This makes the mounting process easier and makes the viewing more successful.