Learn More
Alkali Scientific ASI Supreme™ Frosted Glass Microscope Slides, Positive Charged for Adhesion, 72/cs
SDP
Catalog No. 502389570
Encompass_Preferred
ASI™ Supreme Frosted Glass Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched. These microscope slides are positively charged, to help in holding thin or powdery samples and make them adhere to the slide. This makes the mounting process easier and makes the viewing more successful.
Catalog No. 50-238-9570 Supplier Alkali Scientific Supplier No. SM2576O
ASI™ Supreme Frosted Glass Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched. These microscope slides are positively charged, to help in holding thin or powdery samples and make them adhere to the slide. This makes the mounting process easier and makes the viewing more successful.