Learn More
Alkali Scientific ASI Supreme™ Plain Surface Non-Charged Microscope Slides, Frosted White Writing Area, 72/pk
SDP
Catalog No. 502335779
Encompass_Preferred
ASI™ Scientific Supreme™ Plain Surface Non-Charged Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched.
Catalog No. 50-233-5779 Supplier Alkali Scientific Supplier No. SM2550
ASI™ Scientific Supreme™ Plain Surface Non-Charged Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched.