Learn More
Alkali Scientific CellPro™ Trypsin-EDTA Solution 0.25% in HBSS, with Ca/Mg, 100 mL
SDP
Catalog No. 502336062
Encompass_Preferred
This perfect blend of trypsin (0.25%) and EDTA (0.53mM) is combined in calcium-free and magnesium-free Hanks Balanced Salt Solution along with phenol red. It has been shown to be a safe and effective means of dissociating a wide variety of cells in culture. Virus Screened and mycoplasma tested. Lot number traceability. Sterile-filtered. Porcine parvovirus screened. Ships on dry ice. Store frozen at -20 to -10°C.
Catalog No. 50-233-6062 Supplier Alkali Scientific Supplier No. FB85
This perfect blend of trypsin (0.25%) and EDTA (0.53mM) is combined in calcium-free and magnesium-free Hanks Balanced Salt Solution along with phenol red. It has been shown to be a safe and effective means of dissociating a wide variety of cells in culture. Virus Screened and mycoplasma tested. Lot number traceability. Sterile-filtered. Porcine parvovirus screened. Ships on dry ice. Store frozen at -20 to -10°C.