Learn More
Enzo Life Sciences Asialo-GM2 (0.1mg). CAS: 35960-33-9
SDP
Supplier:  Enzo Life Sciences ALX302015MC01
Encompass_Preferred
A major neural ganglioside in Tay Sachs and Sandhoff diseases. Alternative name: Asialoganglioside-GM2, Ceramide trihexoside, Gangliotriosylceramide. Formulation: Lyophilized. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-200-8656