Learn More
Cayman Chemical AsIalo GnglIosIde GM2 100ug
SDP
Catalog No. 502934968
Encompass_Preferred
A sphingolipid with variable fatty acyl chain lengths; a ganglioside lacking the sialic acid residue; levels are low-to-undetectable in normal human brain, but it accumulates in the brain of patients with Tay-Sachs and Sandhoff disease; binds to various bacteria, including Pseudomonas isolates derived from cystic fibrosis patients
Catalog No. 50-293-4968 Supplier Cayman Chemical Supplier No. 24629100 UG
A sphingolipid with variable fatty acyl chain lengths; a ganglioside lacking the sialic acid residue; levels are low-to-undetectable in normal human brain, but it accumulates in the brain of patients with Tay-Sachs and Sandhoff disease; binds to various bacteria, including Pseudomonas isolates derived from cystic fibrosis patients