Learn More
Santa Cruz Biotechnology Asialoglycoprotein Receptor 1/2/ASGR1/ASGR2 Antibody (E-1) 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC166633
Encompass
Asialoglycoprotein Receptor 1/2/ASGR1/ASGR2 Antibody (E-1) is a mouse monoclonal IgG2b κ Asialoglycoprotein Receptor 1/2/ASGR1/ASGR2 antibody, cited in 3 publications, provided at 200 µg/mlraised against amino acids 1-291 representing full length ASGPR1 of human originAsialoglycoprotein Receptor 1/2/ASGR1/ASGR2 Antibody (E-1) is recommended for detection of ASGPR1, ASGPR2 and MASGP-BP of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC3900880