Learn More
Sino Biological Asialoglycoprotein Receptor / ASGR1 Antibody (APC), Mouse MAb 25Tests
Supplier:  Sino Biological 10773MM02A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Asialoglycoprotein Receptor / ASGR1 (rh Asialoglycoprotein Receptor / ASGR1; Catalog#10773-H07H; P07306; Gln62-Leu291) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-6346