Learn More
Sino Biological Recombinant Mouse ASGR1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50083M07HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Ser60-Asn284) of mouse ASGPR1 (NP_033844.1) was expressed with a N-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-4834