Learn More
Sino Biological Recombinant Human ASGR1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10773H07HB100
Encompass_Preferred
A DNA sequence encoding the human ASGR1 isoform 1 (P07306) (Gln62-Leu291) was expressed with a polyhistidine tag at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6165