Learn More
Cayman Chemical AsIaloglycoproteIn RECPTR 1
SDP
Catalog No. 502916427
Encompass_Preferred
A p300 and CBP inhibitor binds to the p300 and CBP bromodomains (Kds 167 and 151 nM respectively) selective for p300 and CBP over BRD4 as well as a panel of 104 nuclear receptors and ion channels and a panel of 32 enzymes at 10 UM displaces acetylated histones from CBP in a cell-free assay (IC50 170 nM) reduces colony formation and increases differentiation of primary murine leukemic blasts and delays disease initiation following leukemic blast transplantation into sub-lethally irradiated mice when used at concentrations of 5 and 10 UM
Catalog No. 50-291-6427 Supplier Cayman Chemical Supplier No. 38095200 UL
A p300 and CBP inhibitor binds to the p300 and CBP bromodomains (Kds 167 and 151 nM respectively) selective for p300 and CBP over BRD4 as well as a panel of 104 nuclear receptors and ion channels and a panel of 32 enzymes at 10 UM displaces acetylated histones from CBP in a cell-free assay (IC50 170 nM) reduces colony formation and increases differentiation of primary murine leukemic blasts and delays disease initiation following leukemic blast transplantation into sub-lethally irradiated mice when used at concentrations of 5 and 10 UM