Learn More
Cayman Chemical AsIaloglycoproteIn RECPTR 1
SDP
Supplier:  Cayman Chemical 38064100 UG
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged asialoglycoprotein receptor 1 extracellular domain expressed in HEK293 cells • Amino acids: 62-291 • MW: 28.8 kDa
Catalog No. 50-291-8082