Learn More
Cayman Chemical AsIaloglycoproteIn RECPTR 1
SDP
Supplier:  Cayman Chemical 38103100 UL
Encompass_Preferred
A σ1 receptor agonist inhibits reductions in mitochondrial oxygen consumption and lipid peroxidation induced by AB (25-35) in isolated mouse hippocampus at 1 mg/kg improves deficits in novel object recognition induced by AB (25-35) in mice as well as reduces tau phosphorylation and AB (1-42) levels in the hippocampus in the same model at 1 mg/kg increases the latency to fall in the rotarod test reduces gait impairments and reduces increases in the mean acoustic startle response in an Mecp2-/- mouse model of Rett syndrome at 30 mg/kg per day
Catalog No. 50-291-6334