Learn More
Cayman Chemical Asialoglycoprotein Receptor 1 Extracellular Domain (human, recombinant), Purity: ≥90% estimated by SDS-PAGE, 1 mg
SDP
Catalog No. 502916630
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged asialoglycoprotein receptor 1 extracellular domain expressed in HEK293 cells • Amino acids: 62-291 • MW: 28.8 kDa
Catalog No. 50-291-6630 Supplier Cayman Chemical Supplier No. 380641 MG
Source: Recombinant human C-terminal His-tagged asialoglycoprotein receptor 1 extracellular domain expressed in HEK293 cells • Amino acids: 62-291 • MW: 28.8 kDa