Learn More
Cayman Chemical Asialoglycoprotein Receptor 1 Rabbit Monoclonal Antibody (Clone 021), 1 mL
SDP
Catalog No. 502918055
Encompass_Preferred
Immunogen: Recombinant human asialoglycoprotein receptor 1 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Asialoglycoprotein receptor 1 • Applications: ELISA (capture), IP, WB • MW = 33 kDa
Catalog No. 50-291-8055 Supplier Cayman Chemical Supplier No. 381031 ML
Immunogen: Recombinant human asialoglycoprotein receptor 1 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Asialoglycoprotein receptor 1 • Applications: ELISA (capture), IP, WB • MW = 33 kDa