Learn More
Cayman Chemical Asiatic Acid, Purity: ≥95%, CAS: 464-92-6, 100 mg
SDP
Catalog No. 502954740
Encompass_Preferred
A natural triterpene which stimulates wound healing; induces cell cycle arrest and apoptosis in breast cancer cells (IC50 = 5.9 µM for MCF-7 cells); blocks angiogenesis in glioblastomas; down regulates BACE1 while increasing ADAM10 maturation in primary rat cortical neurons; protects against neuronal damage and cognitive defects resulting from glutamate administration in vivo in mice
Catalog No. 50-295-4740 Supplier Cayman Chemical Supplier No. 11818100 MG
A natural triterpene which stimulates wound healing; induces cell cycle arrest and apoptosis in breast cancer cells (IC50 = 5.9 µM for MCF-7 cells); blocks angiogenesis in glioblastomas; down regulates BACE1 while increasing ADAM10 maturation in primary rat cortical neurons; protects against neuronal damage and cognitive defects resulting from glutamate administration in vivo in mice