Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asiatic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014231 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4231 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4231 Supplier MilliporeSigma Supelco Supplier No. 8977310MG
Add to Cart
Add to Cart
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 464-92-6
Physical Form Neat
Linear Formula C30H48O5
MDL Number MFCD00238541
Melting Point 325°C to 330°C
Synonym 2 alpha,23-Dihydroxyursolic acid; Dammarolic acid; NSC 166063
Type Analytical Standard
Quantity 10 mg
Formula Weight 488.70
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.