Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asiatic acid, 97%

Catalog No. AAH60149ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60149ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60149ME Supplier Thermo Scientific Chemicals Supplier No. H60149ME
Add to Cart

CAS: 464-92-6 | C30H48O5 | 488.709 g/mol

Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen phosphorylase inhibitors and and hepatoprotectants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen p
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-92-6
Molecular Formula C30H48O5
Molecular Weight (g/mol) 488.709
MDL Number MFCD00238541
InChI Key JXSVIVRDWWRQRT-YLPWNERUSA-N
Synonym Dammarolic acid
PubChem CID 129316838
IUPAC Name (4aS,6aS,6aS,6bR,8aS,9R,12aR,14bR)-10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid
SMILES CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CC(C(C5(C)CO)O)O)C)C)C2C1C)C)C(=O)O

Specifications

Melting Point 325°C to 330°C
Quantity 500 mg
Solubility Information Soluble in dimethyl sulfoxide,ethanol and dimethylformamide. Sparingly soluble in water
Formula Weight 488.7
Percent Purity 97%
Chemical Name or Material Asiatic acid
RTECSNumber YU9580000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.