Learn More
Cayman Chemical Asiaticoside, Purity: ≥95%, CAS: 16830-15-2, 50 mg
SDP
Catalog No. 502952836
Encompass_Preferred
The main saponin constituent of C. asiatica that has been shown to improve wound healing including increasing collagen synthesis and elevating antioxidant levels in various normal and diabetic wound models
Catalog No. 50-295-2836 Supplier Cayman Chemical Supplier No. 1181950 MG
The main saponin constituent of C. asiatica that has been shown to improve wound healing including increasing collagen synthesis and elevating antioxidant levels in various normal and diabetic wound models