Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Asiaticoside, 92+%

Catalog No. AC461670010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461670010 1 g
AC461670050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461670010 Supplier Thermo Scientific Chemicals Supplier No. 461670010

CAS: 16830-15-2 | C48H78O19 | 959.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16830-15-2
Molecular Formula C48H78O19
Molecular Weight (g/mol) 959.13
InChI Key WYQVAPGDARQUBT-NNUNPJGWSA-N
IUPAC Name 6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}methyl)-3,4,5-trihydroxyoxan-2-yl (1S,2R,4aS,6aS,6bR,9R,10S,11R,12aR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate
SMILES C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CCC4[C@@]5(C)C[C@@H](O)[C@@H](O)[C@@](C)(CO)C5CC[C@@]34C)[C@@H]2[C@H]1C)C(=O)OC1OC(COC2OC(CO)C(OC3OC(C)C(O)C(O)C3O)C(O)C2O)C(O)C(O)C1O

Specifications

Melting Point 230.0°C to 233.0°C
Assay Percent Range 92.0 % min. (HPLC)
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in warmed methanol
Formula Weight 959.12
Chemical Name or Material Asiaticoside
EINECSNumber 240-851-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.