Learn More
Cayman Chemical Asiaticoside B, Purity: ≥98%, CAS: 125265-68-1, 10 mg
SDP
Catalog No. 502947831
Encompass_Preferred
A triterpenoid glycoside; cytotoxic to HepG2 and MCF-7 cancer cells (IC50s = 9.74 and 8.32 µM, respectively)
Catalog No. 50-294-7831 Supplier Cayman Chemical Supplier No. 3342710 MG
A triterpenoid glycoside; cytotoxic to HepG2 and MCF-7 cancer cells (IC50s = 9.74 and 8.32 µM, respectively)