Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asiaticoside B | 125265-68-1 | 5 MG
SDP

Catalog No. 5000420022
Encompass_Preferred

Asiaticoside B is a 9,19-cycloartane triterpene glycoside found in Actaea asiatica. It exhibits cytotoxicity against HepG2 and MCF-7 cancer cells with IC50 values of 9.74 μM and 8.32 μM, respectively.

  • Purity: 99.46%
  • Molecular weight: 975.12
  • Formula: C48H78O20
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants (Compositae, Tagetes lucida Cav., Umbelliferae)
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility (in vitro): DMSO: 100 mg/mL (102.55 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Catalog No. 50-004-20022 Supplier Medchemexpress LLC Supplier No. HYN50525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Asiaticoside B is a 9,19-cycloartane triterpene glycoside found in Actaea asiatica. It exhibits cytotoxicity against HepG2 and MCF-7 cancer cells with IC50 values of 9.74 μM and 8.32 μM, respectively.

  • Purity: 99.46%
  • Molecular weight: 975.12
  • Formula: C48H78O20
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants (Compositae, Tagetes lucida Cav., Umbelliferae)
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility (in vitro): DMSO: 100 mg/mL (102.55 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.