Learn More
Medchemexpress LLC Asimadoline hydrochloride | 185951-07-9 | 99.6% | 451.00 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY107384A
Encompass_Preferred
Asimadoline hydrochloride is an orally active, selective, and peripherally active κ-opioid agonist. It demonstrates low permeability across the blood-brain barrier and exhibits peripheral anti-inflammatory actions. Research indicates its potential for treating irritable bowel syndrome (IBS) and ameliorating allodynia in diabetic rats.
  • Orally active and selective κ-opioid agonist
  • Low permeability across the blood-brain barrier
  • Exhibits peripheral anti-inflammatory actions
  • Potential for irritable bowel syndrome treatment
  • Ameliorates allodynia in diabetic rats
  • Rapid metabolism across species
  • Highly selective for κ-opioid receptors
Catalog No. 50-003-65747