Learn More
Medchemexpress LLC Asimadoline hydrochloride | 185951-07-9 | MFCD28987467 | 99.8% | 451.00 | C27H31ClN2O2 | 25 MG
SDP
Catalog No. 5000239923
Encompass_Preferred
Asimadoline hydrochloride is a research-grade, peripherally selective κ-opioid receptor agonist supplied as the hydrochloride salt for preclinical pharmacology and biochemical studies. It is intended for laboratory research use only and is not for human or veterinary applications. The compound is offered in milligram-scale vials and in ready-to-use DMSO solutions for assay development and in vivo experimentation where permitted.
  • Peripherally selective κ-opioid receptor agonist for pharmacology studies.
  • Provided as the hydrochloride salt for improved solubility.
  • Available in small vial sizes and DMSO solutions for ready use.
  • High purity suitable for analytical and biological assays.
  • Not for human or veterinary use; research use only.
Catalog No. 50-002-39923 Supplier Medchemexpress LLC Supplier No. HY107384A25MG
Asimadoline hydrochloride is a research-grade, peripherally selective κ-opioid receptor agonist supplied as the hydrochloride salt for preclinical pharmacology and biochemical studies. It is intended for laboratory research use only and is not for human or veterinary applications. The compound is offered in milligram-scale vials and in ready-to-use DMSO solutions for assay development and in vivo experimentation where permitted.
  • Peripherally selective κ-opioid receptor agonist for pharmacology studies.
  • Provided as the hydrochloride salt for improved solubility.
  • Available in small vial sizes and DMSO solutions for ready use.
  • High purity suitable for analytical and biological assays.
  • Not for human or veterinary use; research use only.