Learn More
Arlington Scientific ASiManager-AT Agglutination Analyzer
SDP
Catalog No. 22415201 Shop All Arlington Scientific Products
Change view
Click to view available options
Throughput:
450 Tests/hr.
Catalog No. Throughput
22-415-201 450 Tests/hr.
1 options
Catalog No. 22-415-201 Supplier Arlington Scientific Supplier No. 2400000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
World's first semi-automated nontreponemal RPR analyzer
An integrated digital particle analyzer designed to objectively interpret the Arlington Scientific RPR Card Test for Syphilis, other ASI serology tests, and the SureVue RPR.
  • Provides standardized test interpretation and initial predictive titer analysis
  • Eliminates subjectivity and brings data management capability to serology
  • Reads up to 450 tests per hour
  • Provides storage, retrieval, and transmittal of test results
For in vitro diagnostic use only
Specifications
Throughput 450 Tests/hr.
Calibration Yes
Quantity 450 Tests