Learn More
New England Biolabs, Inc. AsiSI – 2500 units
SDP
Catalog No. 50812373
Encompass
A restriction endonuclease that recognizes the sequence GCG_AT^CGC.
Catalog No. 50-812-373 Supplier New England Biolabs, Inc. Supplier No. R0630L
A restriction endonuclease that recognizes the sequence GCG_AT^CGC.