Learn More
Eurofins Discoverx ASK1, active, 250 µg. (14-606M)
Supplier:  Eurofins Discoverx 14606M
Encompass
N-terminal GST and C-terminal 6xHis-tagged recombinant, human ASK1 amino acids 649-946. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9042