Learn More
Medchemexpress LLC HY-133554 5mg Medchemexpress, ASK1-IN-1 CAS:2411382-24-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1335545MG
Encompass_Preferred
Medchemexpress, HY-133554 5mg ASK1-IN-1 CAS:2411382-24-4 ASK1-IN-1 is a CNS-penetrant ASK1 (apoptosis signal-regulating kinase 1) inhibitor, with good potency (cell IC50=138 nM; Biochemical IC50=21 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1346