Learn More
Medchemexpress LLC Ask1-in-2 | 2541792-70-3 | MFCD11217007 | 99.6% | C19H17FN6O | 10MG
SDP
Catalog No. 5000258948
Encompass_Preferred
ASK1-IN-2 is a small-molecule, orally active inhibitor of apoptosis signal-regulating kinase 1 (ASK1) with a reported biochemical IC50 of 32.8 nM. Supplied as a high-purity research chemical (CAS 2541792-70-3; formula C19H17FN6O), it is available as a solid powder and as a 10 mM solution in DMSO for use in in vitro and in vivo preclinical studies, including research into inflammatory conditions such as ulcerative colitis.
  • Potent ASK1 inhibition (IC50 32.8 nM).
  • Orally active small molecule suitable for in vivo studies.
  • High purity (99.6%).
  • Available as solid powder and as 10 mM solution in DMSO.
  • Suitable for research into inflammatory diseases such as ulcerative colitis.
  • Provided with safety data sheet and product documentation.
Catalog No. 50-002-58948 Supplier Medchemexpress LLC Supplier No. HY13196910MG
ASK1-IN-2 is a small-molecule, orally active inhibitor of apoptosis signal-regulating kinase 1 (ASK1) with a reported biochemical IC50 of 32.8 nM. Supplied as a high-purity research chemical (CAS 2541792-70-3; formula C19H17FN6O), it is available as a solid powder and as a 10 mM solution in DMSO for use in in vitro and in vivo preclinical studies, including research into inflammatory conditions such as ulcerative colitis.
  • Potent ASK1 inhibition (IC50 32.8 nM).
  • Orally active small molecule suitable for in vivo studies.
  • High purity (99.6%).
  • Available as solid powder and as 10 mM solution in DMSO.
  • Suitable for research into inflammatory diseases such as ulcerative colitis.
  • Provided with safety data sheet and product documentation.