Learn More
Aviva Systems Biology Corp Asl Antibody - C-terminal region (ARP61235_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61235_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Asl. It was validated on Western Blot. . Alternate names: 2510006M18RikApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-310