Learn More
Medchemexpress LLC HY-129239 5mg Medchemexpress, ASLAN003 CAS:1035688-66-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1292395MG
Encompass_Preferred
Medchemexpress, HY-129239 5mg ASLAN003 CAS:1035688-66-4 ASLAN003 is an orally active and potent inhibitor of DHODH (Human Dihydroorotate Dehydrogenase) which has the potential to be a first-in-class candidate in AML (Acute Myelogenous Leukemia). ASLAN003 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9801