Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ASN007 benzenesulfonate | 2055597-39-0 | 99.7% | 632.11 | C28H31ClFN7O5S | 5 MG
SDP

Catalog No. 5000222393
Encompass_Preferred

ASN007 benzenesulfonate is the benzenesulfonate salt of ASN007, a potent ERK1/2 inhibitor used for preclinical research into MAPK/ERK pathway biology and cancers driven by RAS mutations. It is supplied as a high-purity research reagent for in vitro and in vivo studies.

  • Potent ERK1/2 inhibition with low single-digit nanomolar IC50 values.
  • Suitable for research of RAS- and MAPK/ERK-driven cancer models.
  • Benzenesulfonate salt form for improved handling and stability.
  • High purity (≈99.7%) suitable for biological studies.
  • Molecular weight 632.11 and formula C28H31ClFN7O5S.
  • Available in small reagent pack sizes for dose-response and pilot studies.

Catalog No. 50-002-22393 Supplier Medchemexpress LLC Supplier No. HY136579A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ASN007 benzenesulfonate is the benzenesulfonate salt of ASN007, a potent ERK1/2 inhibitor used for preclinical research into MAPK/ERK pathway biology and cancers driven by RAS mutations. It is supplied as a high-purity research reagent for in vitro and in vivo studies.

  • Potent ERK1/2 inhibition with low single-digit nanomolar IC50 values.
  • Suitable for research of RAS- and MAPK/ERK-driven cancer models.
  • Benzenesulfonate salt form for improved handling and stability.
  • High purity (≈99.7%) suitable for biological studies.
  • Molecular weight 632.11 and formula C28H31ClFN7O5S.
  • Available in small reagent pack sizes for dose-response and pilot studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.