Learn More
Medchemexpress LLC Asn007 benzenesulfonate | 2055597-39-0 | 99.7% | C28H31ClFN7O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY136579A10MG
Encompass_Preferred
ASN007 benzenesulfonate is the benzenesulfonate salt form of ASN007, a small-molecule ERK inhibitor supplied as a solid for research use. It is intended for in vitro and preclinical studies investigating ERK signaling and oncology-related pathways.
  • Suitable for in vitro and preclinical assays.
  • High reported purity supports biochemical testing.
  • Soluble in DMSO and water with recommended handling.
  • Stable when stored at -20°C, desiccated.
  • Provided in small milligram quantities for screening and profiling.
Catalog No. 50-002-60856