Learn More
Medchemexpress LLC (Asn10,Leu11,D-Trp12)-pTH-Related Protein (7-34) amide (human, mouse, rat) | 129622-68-0 | 99.3% | 3478.06 | 5 MG
SDP
Catalog No. 5000397171
Encompass_Preferred
(Asn10,Leu11,D-Trp12)-pTH-Related Protein (7-34) amide (human, mouse, rat) is a potent PTH-1R antagonist, showing no partial agonist activity. It completely blocks the antioxidant effect of PTHrP(1-37) in mouse osteoblast MC3T3-E1 and primary human osteoblasts, confirming PTH1R activation as the main signaling mechanism for the antioxidant effect of PTHrP(1-37).
  • Potent PTH-1R antagonist
  • Does not display any partial agonist activity
  • Blocks antioxidant effect of PTHrP(1-37) in osteoblast cells
  • Relevant for research on PTH1R signaling
Catalog No. 50-003-97171 Supplier Medchemexpress LLC Supplier No. HYP43855MG
(Asn10,Leu11,D-Trp12)-pTH-Related Protein (7-34) amide (human, mouse, rat) is a potent PTH-1R antagonist, showing no partial agonist activity. It completely blocks the antioxidant effect of PTHrP(1-37) in mouse osteoblast MC3T3-E1 and primary human osteoblasts, confirming PTH1R activation as the main signaling mechanism for the antioxidant effect of PTHrP(1-37).
  • Potent PTH-1R antagonist
  • Does not display any partial agonist activity
  • Blocks antioxidant effect of PTHrP(1-37) in osteoblast cells
  • Relevant for research on PTH1R signaling