Learn More
Medchemexpress LLC (Asn10,Leu11,D-Trp12)-pTH-Related Protein (7-34) amide (human, mouse, rat) | 129622-68-0 | 99.32% | 3478.06 | 10 MG
SDP
Catalog No. 5000396735
Encompass_Preferred
Asn10,Leu11,D-Trp12-pTH-Related Protein (7-34) amide (human, mouse, rat) is a potent PTH-1R antagonist that does not display any partial agonist activity. This product completely blocks the antioxidant effect of PTHrP(1-37) in mouse osteoblast MC3T3-E1 and primary human osteoblasts, confirming PTH1R activation as the main signaling mechanism for the antioxidant effect of PTHrP(1-37).
  • Potent PTH-1R antagonist
  • Does not display partial agonist activity
  • Blocks antioxidant effect of PTHrP(1-37) in osteoblast cells
  • For research use only
Catalog No. 50-003-96735 Supplier Medchemexpress LLC Supplier No. HYP438510MG
Asn10,Leu11,D-Trp12-pTH-Related Protein (7-34) amide (human, mouse, rat) is a potent PTH-1R antagonist that does not display any partial agonist activity. This product completely blocks the antioxidant effect of PTHrP(1-37) in mouse osteoblast MC3T3-E1 and primary human osteoblasts, confirming PTH1R activation as the main signaling mechanism for the antioxidant effect of PTHrP(1-37).
  • Potent PTH-1R antagonist
  • Does not display partial agonist activity
  • Blocks antioxidant effect of PTHrP(1-37) in osteoblast cells
  • For research use only