Learn More
Aviva Systems Biology Corp ASNA1 Antibody - C-terminal region (ARP60201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASNA1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ASNA1. Alternate names: ARSA-I, ARSA1, MGC3821, GET3, TRC40, ASNA-I, hASNA-IApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-974