Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sure-Vue™ ASO Latex Test Kit
SDP

Catalog No. p-7222290 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-105 50 Tests
23-038-106 100 Tests
23-038-107 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-038-105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Sure-Vue ASO Latex Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of Antistreptolysin-O (ASO) antibodies in human serum.

  • The ASO Slide Test is a latex slide agglutination assay for the qualitative and semiquantitative detection of antistreptolysin-O (ASO) antibodies in human serum.
  • No initial dilution of patient samples is required for this test.
  • These materials are intended to be acquired, possessed and used only by health professionals.

Specifications

Content And Storage 2°C to 8°C
Control Sets Yes
CPT Code 86060
CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Anti-Streptolysin O (ASO)
Detection Method Agglutination
Sample Type Human Serum
Sensitivity 200 IU/mL
Disposable Single-use
Includes Latex reagent, controls, cards, and applicators
Quantity 50 Tests
Sample Size 30 μL
Specificity >95%
State at Room Temperature Liquid
Sterility Non-sterile
Test Time 2 Minutes
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.