Learn More
Sigma Aldrich Fine Chemicals Biosciences Soy Phospholipids, 50 g
Catalog No. NC1292240
Encompass
Asolectin comprises roughly equal proportions of lecithin, cephalin and phosphatidylinositol along with minor amounts of other phospholipids and polar lipids. Good source of fatty acids: about 24% saturated fatty acids, 14% mono-unsaturated and 62% poly-unsaturated fatty acids. For the reconstitution of vesicles in membrane studies; review about the swelling of soybean phospholipids in water and their function as oil-in-water emulsifiers.
Catalog No. NC1292240 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1114550G
Add to Cart
Add to Cart
Asolectin comprises roughly equal proportions of lecithin, cephalin and phosphatidylinositol along with minor amounts of other phospholipids and polar lipids. Good source of fatty acids: about 24% saturated fatty acids, 14% mono-unsaturated and 62% poly-unsaturated fatty acids. For the reconstitution of vesicles in membrane studies; review about the swelling of soybean phospholipids in water and their function as oil-in-water emulsifiers.