Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asoxime chloride | 34433-31-3 | MFCD00869535 | 98.5% | 359.21 g/mol | C14H16Cl2N4O3 | 5 MG
SDP

Catalog No. 5000239947
Encompass_Preferred

Asoxime dichloride (HI-6) is a research-grade acetylcholine receptor antagonist used in pharmacology and neuroscience studies. It interacts with nicotinic receptors, including α7 nAChR, and is used for receptor modulation studies, immunology research, and as a defined reference material in analytical workflows.

  • CAS number 34433-31-3.
  • Molecular formula C14H16Cl2N4O3 and molecular weight 359.21 g/mol.
  • High purity, listed as approximately 98.5%.
  • Available in small research quantities (mg-scale) for laboratory use.
  • Supplied as the dichloride salt suitable for receptor pharmacology and immunological assays.

Catalog No. 50-002-39947 Supplier Medchemexpress LLC Supplier No. HY106901A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Asoxime dichloride (HI-6) is a research-grade acetylcholine receptor antagonist used in pharmacology and neuroscience studies. It interacts with nicotinic receptors, including α7 nAChR, and is used for receptor modulation studies, immunology research, and as a defined reference material in analytical workflows.

  • CAS number 34433-31-3.
  • Molecular formula C14H16Cl2N4O3 and molecular weight 359.21 g/mol.
  • High purity, listed as approximately 98.5%.
  • Available in small research quantities (mg-scale) for laboratory use.
  • Supplied as the dichloride salt suitable for receptor pharmacology and immunological assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.