Learn More
Cayman Chemical AsoxImechlorIde 10mg
SDP
Catalog No. 502980258
Encompass_Preferred
An asymmetric bis-pyridinium aldoxime that functions as an AChE reactivator through nucleophilic attack of oximate anions on organophosphate-AChE conjugates; demonstrates therapeutic activity in experimental models of organophosphate poisoning
Catalog No. 50-298-0258 Supplier Cayman Chemical Supplier No. 1860710 MG
An asymmetric bis-pyridinium aldoxime that functions as an AChE reactivator through nucleophilic attack of oximate anions on organophosphate-AChE conjugates; demonstrates therapeutic activity in experimental models of organophosphate poisoning